missing translation for 'onlineSavingsMsg'
Learn More
4'-(Bromomethyl)biphenyl-2-carbonitrile, 98%, Thermo Scientific Chemicals Product Code.: 15421618

4'-(Bromomethyl)biphenyl-2-carbonitrile, 98%, Thermo Scientific Chemicals

Product Code. 15421618
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15421618

Brand: Thermo Scientific Alfa Aesar H55394.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114772-54-2
Molecular Formula C14H10BrN
Molecular Weight (g/mol) 272.145
MDL Number MFCD00671503
InChI Key LFFIEVAMVPCZNA-UHFFFAOYSA-N
Synonym 4'-bromomethyl-2-cyanobiphenyl, 2-4-bromomethyl phenyl benzonitrile, 2-cyano-4'-bromomethylbiphenyl, 4'-bromomethyl-1,1'-biphenyl-2-carbonitrile, 2-4-bromo-methylphenyl benzonitrile, 4'-bromomethyl-2-biphenylcarbonitrile, 4'-bromomethylbiphenyl-2-carbonitrile, 2'-cyano-4-bromomethylbiphenyl, 4-2-cyanophenyl benzyl bromide, unii-ui5jr3k10fShow MoreShow Less
PubChem CID 1501912
IUPAC Name 2-[4-(bromomethyl)phenyl]benzonitrile
SMILES C1=CC=C(C(=C1)C#N)C2=CC=C(C=C2)CBr

Specifications

Melting Point 125°C to 128°C
Quantity 25 g
UN Number UN3439
Formula Weight 272.14
Percent Purity 98%
Chemical Name or Material 4'-(Bromomethyl)biphenyl-2-carbonitrile

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR18G013 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH55394.14,H55394.06
Lot NumberY04F001 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH55394.14,H55394.06
Lot NumberR25H057 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH55394.14,H55394.06
Lot NumberQ29H039 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH55394.14,H55394.06
Lot NumberZ28G014 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH55394.14,H55394.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4'-(Bromomethyl)biphenyl-2-carbonitrile, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.