missing translation for 'onlineSavingsMsg'
Learn More
4-Chloro-1,8-naphthalic anhydride, 94%, Thermo Scientific Chemicals Product Code.: 11397465

4-Chloro-1,8-naphthalic anhydride, 94%, Thermo Scientific Chemicals

Product Code. 11397465
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11397465

Brand: Thermo Scientific Alfa Aesar L05508.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-Chloro-1,8-naphthalic anhydride is used in dyes and pigments. It is also used in agrochemical, pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-1,8-naphthalic anhydride is used in dyes and pigments. It is also used in agrochemical, pharmaceutical.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store at 4°C. Store away from moisture. Incompatible with oxidizing agents, water, Strong bases, Strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4053-08-1
Molecular Formula C12H5ClO3
Molecular Weight (g/mol) 232.62
MDL Number MFCD00006928
InChI Key UJEUBSWHCGDJQU-UHFFFAOYSA-N
IUPAC Name 10-chloro-3-oxatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-2,4-dione

Specifications

Melting Point 208°C to 214°C
Quantity 500 g
Beilstein 178072
Sensitivity Moisture sensitive
Synonym 1,2:3,4-di-o-isopropylidene-d-galactose, 1,2:3,4-di-o-isopropylidene-, a-d-galactopyranose, 1,2:3,4-bis-o-1-methylethylidene-, 1s,6r,9s-4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo 7.3.0.0 2 ,? dodecan-8-yl methanol
Solubility Information Hydrolyzes in water.
SMILES ClC1=C2C=CC=C3C(=O)OC(=O)C(C=C1)=C23
PubChem CID 70793572
Formula Weight 232.63
Percent Purity 94%
Chemical Name or Material 4-Chloro-1,8-naphthalic anhydride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU22I018 Certificate TypeCertificate of Analysis Date22/09/2022 Catalog NumberL05508.22,L05508.14,L05508.36
Lot NumberT07I015 Certificate TypeCertificate of Analysis Date19/07/2022 Catalog NumberL05508.22,L05508.14,L05508.36
Lot Number5005N16Q Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberL05508.22,L05508.14,L05508.36
Lot Number5003J29P Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL05508.22,L05508.14,L05508.36
Lot Number5020N01N Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL05508.22,L05508.14,L05508.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Chloro-1,8-naphthalic anhydride, 94%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.