missing translation for 'onlineSavingsMsg'
Learn More
4-Chloro-7-nitrobenzofurazan, 98%, Thermo Scientific Chemicals Product Code.: 11442207

4-Chloro-7-nitrobenzofurazan, 98%, Thermo Scientific Chemicals

Product Code. 11442207
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11442207

Brand: Thermo Scientific Alfa Aesar A14165.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-Chloro-7-nitrobenzofurazan is used as a derivatizing reagent for chromatography analysis of amino acids and low molecular weight amines. It is utilized in the preparation of fluorescent phospholipid-derivative, hydroxynaphthofurazan and 4-chloro-7-nitrobenzofurazan- didecanoylphosphatidylethanolamine. It serves as a fluorescent reagent to label free sulfhydryls and N-terminals within proteins and trapping agent for cysteine sulfenic acid. It is a sensitive chromogenic and fluorogenic reagent. It reacts with trans-1-methoxy-3-(trimethylsilyloxy)-1,3-butadiene (Danishefsky's diene) to prepare regioselectively the silyl enol ether. Further, it is used to label peptides, proteins and other biomolecules. In addition it is used in the synthesis of fluorescent phospholipid-derivative, NBD-didecanoylphosphatidylethanolamine, functionalized hydroxynaphthofurazan and 7-nitrobenzofurazan (NBD)-labeled maleimide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-7-nitrobenzofurazan is used as a derivatizing reagent for chromatography analysis of amino acids and low molecular weight amines. It is utilized in the preparation of fluorescent phospholipid-derivative, hydroxynaphthofurazan and 4-chloro-7-nitrobenzofurazan- didecanoylphosphatidylethanolamine. It serves as a fluorescent reagent to label free sulfhydryls and N-terminals within proteins and trapping agent for cysteine sulfenic acid. It is a sensitive chromogenic and fluorogenic reagent. It reacts with trans-1-methoxy-3-(trimethylsilyloxy)-1,3-butadiene (Danishefsky′s diene) to prepare regioselectively the silyl enol ether. Further, it is used to label peptides, proteins and other biomolecules. In addition it is used in the synthesis of fluorescent phospholipid-derivative, NBD-didecanoylphosphatidylethanolamine, functionalized hydroxynaphthofurazan and 7-nitrobenzofurazan (NBD)-labeled maleimide.

Solubility
Soluble in methanol, dimethylsulfoxide, dimethylformamide and chloroform.

Notes
Store in a cool place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10199-89-0
Molecular Formula C6H2ClN3O3
Molecular Weight (g/mol) 199.55
MDL Number MFCD00005808
InChI Key IGHBXJSNZCFXNK-UHFFFAOYSA-N
Synonym 4-chloro-7-nitrobenzofurazan, nbd chloride, 4-chloro-7-nitrobenzo-2-oxa-1,3-diazole, nbd-cl, 7-chloro-4-nitrobenzofurazan, 2,1,3-benzoxadiazole, 4-chloro-7-nitro, nbd-chloride, nbd-c 1, 1-chloro-4-nitrobenzoxadiazole, 4-nitro-7-chlorobenzofurazan
PubChem CID 25043
ChEBI CHEBI:78878
IUPAC Name 4-chloro-7-nitro-2,1,3-benzoxadiazole
SMILES C1=C(C2=NON=C2C(=C1)Cl)[N+](=O)[O-]

Specifications

Melting Point 96°C to 100°C
Quantity 5 g
Beilstein 614212
Solubility Information Soluble in methanol,dimethylsulfoxide,dimethylformamide and chloroform.
Formula Weight 199.56
Percent Purity 99%
Chemical Name or Material 4-Chloro-7-nitrobenzofurazan

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number181181A Certificate TypeCertificate of Analysis Date21/02/2025 Catalog NumberA14165.06,A14165.14
Lot Number3032433 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog NumberA14165.06,A14165.14
Lot Number10241149 Certificate TypeCertificate of Analysis Date02/04/2024 Catalog NumberA14165.06,A14165.14
Lot Number10235316 Certificate TypeCertificate of Analysis Date17/10/2023 Catalog NumberA14165.06,A14165.14
Lot Number10245055 Certificate TypeCertificate of Analysis Date16/12/2022 Catalog NumberA14165.06,A14165.14
compliance-icons
Product Identifier
  • 4-Chloro-7-nitrobenzofurazan
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 4-Chloro-7-nitrobenzofurazan

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Chloro-7-nitrobenzofurazan, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.