missing translation for 'onlineSavingsMsg'
Learn More
4-Chlorobenzoic acid, 99%, Thermo Scientific Chemicals Product Code.: 10357620

4-Chlorobenzoic acid, 99%, Thermo Scientific Chemicals

Product Code. 10357620
250 g, Plastic bottle
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10357620

Brand: Thermo Scientific Acros 159252500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74-11-3
Molecular Formula C7H5ClO2
Molecular Weight (g/mol) 156.57
MDL Number MFCD00002531
InChI Key XRHGYUZYPHTUJZ-UHFFFAOYSA-N
Synonym p-chlorobenzoic acid, benzoic acid, 4-chloro, p-chlorbenzoic acid, chlorodracylic acid, benzoic acid, p-chloro, para-chlorobenzoic acid, 4-chloro-benzoic acid, p-carboxychlorobenzene, acido p-clorobenzoico, acido p-clorobenzoico italianShow MoreShow Less
PubChem CID 6318
ChEBI CHEBI:30747
IUPAC Name 4-chlorobenzoic acid
SMILES C1=CC(=CC=C1C(=O)O)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 240.0°C to 243.0°C
Color White to Yellow
Boiling Point 274.0°C to 276.0°C
Flash Point 238°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula ClC6H4CO2H
Quantity 250 g
Beilstein 09, 340
Merck Index 15, 2126
Solubility Information Solubility in water: 0,08g/L (20°C). Other solubilities: soluble in hot water, freely soluble in alcohol and ether
Formula Weight 156.57
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Chlorobenzoic acid, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0442446 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number159252500,159250010,159250250
Lot NumberA0394307 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number159252500,159250010,159250250
Lot NumberA0442446 Certificate TypeCertificate of Analysis Date10/06/2022 Catalog Number159252500,159250010,159250250
Lot NumberA0434528 Certificate TypeCertificate of Analysis Date15/10/2021 Catalog Number159252500,159250010,159250250
Lot NumberA0406901 Certificate TypeCertificate of Analysis Date28/09/2021 Catalog Number159252500,159250010,159250250
compliance-icons
Product Identifier
  • 4-Chlorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Chlorobenzoic acid, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.