missing translation for 'onlineSavingsMsg'
Learn More
4-Hydroxybenzhydrazide, 98%, Thermo Scientific Chemicals Product Code.: 11400623

4-Hydroxybenzhydrazide, 98%, Thermo Scientific Chemicals

Product Code. 11400623
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11400623

Brand: Thermo Scientific Alfa Aesar A12702.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5351-23-5
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00007605
InChI Key ZMZGIVVRBMFZSG-UHFFFAOYSA-N
Synonym 4-hydroxybenzhydrazide, 4-hydroxybenzoic acid hydrazide, 4-hydroxybenzoic hydrazide, p-hydroxybenzhydrazide, p-hydroxybenzoic acid hydrazide, 4-hydroxybenzoylhydrazine, benzoic acid, 4-hydroxy-, hydrazide, p-hydroxybenzoic hydrazide, p-hydroxybenzoyl hydrazine, 4-hydroxy-benzoic acid hydrazideShow MoreShow Less
PubChem CID 1742
IUPAC Name 4-hydroxybenzohydrazide
SMILES C1=CC(=CC=C1C(=O)NN)O

Specifications

Melting Point ∼267°C (decomposition)
Quantity 25 g
Beilstein 473052
Formula Weight 152.15
Percent Purity 98%
Chemical Name or Material 4-Hydroxybenzhydrazide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10242010 Certificate TypeCertificate of Analysis Date21/07/2022 Catalog NumberA12702.22,A12702.14,A12702.36
Lot Number10238576 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA12702.22,A12702.14,A12702.36
Lot NumberH7156A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12702.22,A12702.14,A12702.36
Lot Number10213031 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12702.22,A12702.14,A12702.36
Lot Number10101284 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12702.22,A12702.14,A12702.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Hydroxybenzhydrazide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.