missing translation for 'onlineSavingsMsg'
Learn More
4-Hydroxynicotinic acid, 98%, Thermo Scientific Chemicals Product Code.: 15474987

4-Hydroxynicotinic acid, 98%, Thermo Scientific Chemicals

Product Code. 15474987
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15474987

Brand: Thermo Scientific Alfa Aesar H32282.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

4-Hydroxynicotinic acid, is used as a chemical synthesis reagent. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxynicotinic acid, is used as a chemical synthesis reagent. It is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in 0.1 M NaOH: 0.1 g/mL.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-70-1
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00040286
InChI Key CHCUBGPSZDGABM-UHFFFAOYSA-N
Synonym 4-hydroxynicotinic acid, 4-hydroxypyridine-3-carboxylic acid, 4-hydroxy-nicotinic acid, 3-pyridinecarboxylic acid, 4-hydroxy, 4-hydroxy-3-pyridinecarboxylic acid, 4-oxo-1,4-dihydropyridine-3-carboxylic acid, 4-oxo-1,4-dihydro-3-pyridinecarboxylic acid, 4-hydroxyl-nicotinic acid, 3-carboxy-4-hydroxypyridine, odhpcaShow MoreShow Less
PubChem CID 69113
IUPAC Name 4-oxo-1H-pyridine-3-carboxylic acid
SMILES OC(=O)C1=CNC=CC1=O

Specifications

Melting Point 256°C to 259°C
Quantity 1 g
Solubility Information Soluble in 0.1 M NaOH: 0.1g/mL.
Formula Weight 139.11
Percent Purity 98%
Chemical Name or Material 4-Hydroxynicotinic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10158550 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH32282.03
Lot Number10166873 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH32282.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Hydroxynicotinic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.