missing translation for 'onlineSavingsMsg'
Learn More
4-Hydroxyphenoxyacetic acid, 98+%, Thermo Scientific Chemicals Product Code.: 11449773

4-Hydroxyphenoxyacetic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11449773
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11449773

Brand: Thermo Scientific Alfa Aesar A12940.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1878-84-8
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.148
MDL Number MFCD00014362
InChI Key PKGWLCZTTHWKIZ-UHFFFAOYSA-N
Synonym 4-hydroxyphenoxyacetic acid, 4-hydroxyphenoxy acetic acid, 2-4-hydroxyphenoxy acetic acid, 4-hydroxypheoxyacetate, acetic acid, 4-hydroxyphenoxy, 4-hydroxy-phenoxy-acetic acid, p-hydroxyphenoxyacetic acid, acetic acid, 2-4-hydroxyphenoxy, acetic acid, p-hydroxyphenoxy, 4-carboxymethoxy phenolShow MoreShow Less
PubChem CID 15881
ChEBI CHEBI:1881
IUPAC Name 2-(4-hydroxyphenoxy)acetic acid
SMILES C1=CC(=CC=C1O)OCC(=O)O

Specifications

Melting Point 153°C to 158°C
Quantity 100 g
Beilstein 1952779
Formula Weight 168.15
Percent Purity ≥98%
Chemical Name or Material 4-Hydroxyphenoxyacetic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10247369 Certificate TypeCertificate of Analysis Date17/07/2023 Catalog NumberA12940.22,A12940,A12940.14,A12940.06
Lot Number10163357 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12940.22,A12940.14,A12940.06
Lot NumberF5058A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12940.22,A12940.14,A12940.06
Lot Number10140854 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12940.22,A12940.14,A12940.06
Lot Number10123880 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12940.22,A12940.14,A12940.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Hydroxyphenoxyacetic acid, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.