Learn More
4-Methyl-3-nitrothiobenzamide, 97%, Thermo Scientific™
£30.94 - £405.00
Chemical Identifiers
CAS | 1233513-42-2 |
---|---|
Molecular Formula | C8H8N2O2S |
Molecular Weight (g/mol) | 196.22 |
MDL Number | MFCD09025715 |
InChI Key | PGQMEJDSPAENNP-UHFFFAOYSA-N |
Synonym | 4-methyl-3-nitrothiobenzamide |
PubChem CID | 45040726 |
IUPAC Name | 4-methyl-3-nitrobenzenecarbothioamide |
SMILES | CC1=C(C=C(C=C1)C(N)=S)[N+]([O-])=O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1233513-42-2 | |
196.22 | |
PGQMEJDSPAENNP-UHFFFAOYSA-N | |
45040726 | |
CC1=C(C=C(C=C1)C(N)=S)[N+]([O-])=O |
C8H8N2O2S | |
MFCD09025715 | |
4-methyl-3-nitrothiobenzamide | |
4-methyl-3-nitrobenzenecarbothioamide |
Specifications
1233513-42-2 | |
MFCD09025715 | |
PGQMEJDSPAENNP-UHFFFAOYSA-N | |
4-methyl-3-nitrobenzenecarbothioamide | |
45040726 | |
97% |
C8H8N2O2S | |
4-methyl-3-nitrothiobenzamide | |
CC1=C(C=C(C=C1)C(N)=S)[N+]([O-])=O | |
196.22 | |
196.22 | |
4-Methyl-3-nitrothiobenzamide |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger