missing translation for 'onlineSavingsMsg'
Learn More
4-Nitrophenyl-2-acetamido-2-deoxy-β-D-glucopyranoside, 99%, Thermo Scientific Chemicals Product Code.: 10299533

4-Nitrophenyl-2-acetamido-2-deoxy-β-D-glucopyranoside, 99%, Thermo Scientific Chemicals

Product Code. 10299533
1 g, Glass bottle
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10299533

Brand: Thermo Scientific Acros 229410010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3459-18-5
Molecular Formula C14H18N2O8
Molecular Weight (g/mol) 342.30
MDL Number MFCD00063696
InChI Key OMRLTNCLYHKQCK-JQXIRTQVNA-N
Synonym 4-nitrophenyl-n-acetyl-beta-d-glucosaminide, 4-nitrophenyl 2-acetamido-2-deoxy-beta-d-glucopyranoside, 4-nitrophenyl-n-acetyl-2-deoxyglucopyranoside, 4'-nitrophenyl-2-acetamido-2-deoxy-beta-glucopyranoside, p-nitrophenyl n-acetyl-beta-d-glucosaminide, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy oxan-3-yl acetamide, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy tetrahydro-2h-pyran-3-yl acetamide, 4-nitrophenyl 2-acetamido-2-deoxy-b-d-glucopyranoside, lec, n-2s,3r,4r,5s,6r-4,5-dihydroxy-6-hydroxymethyl-2-4-nitrophenoxy oxan-3-yl ethanamideShow MoreShow Less
PubChem CID 102416
IUPAC Name N-[(2S,3R,4R,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-nitrophenoxy)oxan-3-yl]acetamide
SMILES CC(=O)N[C@@H]1[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]1OC1=CC=C(C=C1)[N+]([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 210.0°C to 212.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 18, V,11, 102
Specific Rotation Condition −16° (20°C c=0.5,water)
Specific Rotation − 16.00
Solubility Information (0.5% in water) Clear colorless to light yellow
Formula Weight 342.29
Percent Purity 99%
Physical Form Fine Powder
Chemical Name or Material 4-Nitrophenyl-2-acetamido-2-deoxy-β-D-glucopyranoside, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468942 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number229411000,229410010
Lot NumberA0468464 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number229411000,229410010
Lot NumberA0466607 Certificate TypeCertificate of Analysis Date03/10/2024 Catalog Number229411000,229410010
Lot NumberA0464285 Certificate TypeCertificate of Analysis Date15/07/2024 Catalog Number229411000,229410010
Lot NumberA0461868 Certificate TypeCertificate of Analysis Date15/04/2024 Catalog Number229411000,229410010

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Nitrophenyl-2-acetamido-2-deoxy-β-D-glucopyranoside, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.