missing translation for 'onlineSavingsMsg'
Learn More
4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals Product Code.: 10386720

4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals

Product Code. 10386720
100 g, Plastic bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10386720

Brand: Thermo Scientific Acros 128751000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 830-03-5
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.15
MDL Number MFCD00007326
InChI Key QAUUDNIGJSLPSX-UHFFFAOYSA-N
Synonym p-nitrophenyl acetate, p-acetoxynitrobenzene, acetic acid, 4-nitrophenyl ester, p-nitrophenol acetate, acetic acid p-nitrophenyl ester, acetic acid, p-nitrophenyl ester, p-nitrophenyl acetate van, 4-nitrophenyl acetate, unii-i902j0qh9s, acetic acid 4-nitrophenyl esterShow MoreShow Less
PubChem CID 13243
ChEBI CHEBI:82635
SMILES CC(=O)OC1=CC=C(C=C1)[N+]([O-])=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 76.0°C to 79.0°C
Color Green
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula CH3CO2C6H4NO2
Quantity 100 g
Beilstein 06, 233
Fieser 02,297
Solubility Information Solubility in water: insoluble
IUPAC Name 4-nitrophenyl acetate
Formula Weight 181.15
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Nitrophenyl acetate, 97%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468476 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number128751000,128750050,128750250
Lot NumberA0465114 Certificate TypeCertificate of Analysis Date15/08/2024 Catalog Number128751000,128750050,128750250
Lot NumberA0434600 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog Number128751000,128750050,128750250
Lot NumberA0429670 Certificate TypeCertificate of Analysis Date16/11/2023 Catalog Number128751000,128750050,128750250
Lot NumberA0456663 Certificate TypeCertificate of Analysis Date02/10/2023 Catalog Number128751000,128750050,128750250
Nombre del producto
  • 4-Nitrophenyl acetate

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
4-Nitrophenyl acetate, 97%, Thermo Scientific Chemicals >

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado