missing translation for 'onlineSavingsMsg'
Learn More
5-(4-Chlorophenyl)thiazolo[2,3-c]-1,2,4-triazole-3-thiol, 96%, Thermo Scientific™ Product Code.: 15498545

5-(4-Chlorophenyl)thiazolo[2,3-c]-1,2,4-triazole-3-thiol, 96%, Thermo Scientific™

Product Code. 15498545
500 mg
Click to view available options
Quantity:
500 mg
This item is not returnable. View return policy

Product Code. 15498545

Brand: Thermo Scientific Alfa Aesar H33621.ME

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 610276-80-7
Molecular Formula C10H6ClN3S2
MDL Number MFCD06254380
Quantity 500 mg
Synonym 5-4-chlorophenyl 1,3 thiazolo 2,3-c 1,2,4 triazole-3-thiol, 5-4-chlorophenyl-1,2,4 triazolo 3,4-b 1,3 thiazole-3-thiol, 5-4-chlorophenyl thiazolo 2,3-c-1,2,4-triazole-3-thiol, 5-4-chlorophenyl thiazolo 2,3-c 1,2,4 triazole-3-thiol, 5-4-chlorophenyl-2h-1,3 thiazolo 2,3-c 1,2,4 triazole-3-thione, 5-4-chlorophenyl-2h-1,2,4 triazolo 3,4-b 1,3 thiazole-3-thione
InChI Key QSDOIJACNQRZKE-UHFFFAOYSA-N
SMILES ClC1=CC=C(C=C1)C1=CSC2=NNC(=S)N12
IUPAC Name 5-(4-chlorophenyl)-2H-[1,3]thiazolo[2,3-c][1,2,4]triazole-3-thione
Molecular Weight (g/mol) 267.75
PubChem CID 4738267
Formula Weight 267.76
Percent Purity 96%
Chemical Name or Material 5-(4-Chlorophenyl)thiazolo[2,3-c]-1,2,4-triazole-3-thiol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-(4-Chlorophenyl)thiazolo[2,3-c]-1,2,4-triazole-3-thiol, 96%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.