missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-(4-Methylphenyl)thiazolo[2,3-c]-1,2,4-triazole-3-thiol, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Specifications
Specifications
CAS | 500112-74-3 |
Melting Point | 194°C to 195°C |
Molecular Formula | C11H9N3S2 |
MDL Number | MFCD06254379 |
Quantity | 500 mg |
Synonym | 5-4-methylphenyl 1,3 thiazolo 2,3-c 1,2,4 triazole-3-thiol, 5-4-methylphenyl-1,2,4 triazolo 3,4-b 1,3 thiazole-3-thiol, 5-p-tolyl thiazolo 2,3-c 1,2,4 triazole-3-thiol, 5-4-methylphenyl-2h-1,3 thiazolo 2,3-c 1,2,4 triazole-3-thione, 5-4-methylphenyl-2h-1,2,4 triazolo 3,4-b 1,3 thiazole-3-thione, 5-4-methylphenyl thiazolo 2,3-c-1,2,4-triazole-3-thiol, 5-p-tolyl-2h-thiazolo 2,3-c 1,2,4 triazole-3-thione, 5-4-methylphenyl-2h-thiazolo 2,3-c 1,2,4 triazole-3-thione, 5-4-methylphenyl-2h-1,3-thiazolino 2,3-c 1,2,4-triazoline-3-thione, 5-4-methylphenyl 1,3 thiazolo 2,3-c 1,2,4 triazole-3 2h-thione |
InChI Key | LNDYWRMGFKFDTC-UHFFFAOYSA-N |
SMILES | CC1=CC=C(C=C1)C2=CSC3=NNC(=S)N23 |
IUPAC Name | 5-(4-methylphenyl)-2H-[1,3]thiazolo[2,3-c][1,2,4]triazole-3-thione |
Molecular Weight (g/mol) | 247.334 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10168790 | Certificate of Analysis | 07/12/2021 | H33012.ME |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
5-(4-Methylphenyl)thiazolo[2,3-c]-1,2,4-triazole-3-thiol, 96%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction