missing translation for 'onlineSavingsMsg'
Learn More
5,5'-Dibromo-2,2'-bithiophene, 98%, Thermo Scientific Chemicals Product Code.: 15412808

5,5'-Dibromo-2,2'-bithiophene, 98%, Thermo Scientific Chemicals

Product Code. 15412808
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 15412808

Brand: Thermo Scientific Alfa Aesar H56337.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is an active pharmaceutical ingredient and an OLED (organic light-emitting diode) intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical ingredient and an OLED (organic light-emitting diode) intermediate.

Solubility
Slightly soluble in water.

Notes
Recommended storage temperature: 2 - 4°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4805-22-5
Molecular Formula C8H4Br2S2
Molecular Weight (g/mol) 324.048
MDL Number MFCD00219110
InChI Key SXNCMLQAQIGJDO-UHFFFAOYSA-N
Synonym 5,5'-dibromo-2,2'-bithiophene, 2-bromo-5-5-bromothiophen-2-yl thiophene, 5,5-dibromo-2,2-bithiophene, 2,2'-bithiophene, 5,5'-dibromo, 5,5'-bis 2-bromothiophene, 5,5'-dibromo-2,2'bithiophene, 5,5'-dibromo-2,2-bithiophene, maybridge4_002009, acmc-209kc4, ksc491i4rShow MoreShow Less
PubChem CID 638908
IUPAC Name 2-bromo-5-(5-bromothiophen-2-yl)thiophene
SMILES C1=C(SC(=C1)Br)C2=CC=C(S2)Br

Specifications

Melting Point 144°C to 146°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 324.06
Percent Purity 99%
Chemical Name or Material 5,5'-Dibromo-2,2'-bithiophene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberBB160501A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH56337.06,H56337.14
Lot Number81500084 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH56337.06,H56337.14
Lot NumberU27H015 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH56337.06,H56337.14
Lot NumberZ14G053 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH56337.06,H56337.14
Lot Number81302556 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH56337.06,H56337.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5,5'-Dibromo-2,2'-bithiophene, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.