missing translation for 'onlineSavingsMsg'
Learn More
5,6-Dimethylthieno[2,3-d]pyrimidine-4(3H)-thione, 96%, Thermo Scientific™ Product Code.: 15438485

5,6-Dimethylthieno[2,3-d]pyrimidine-4(3H)-thione, 96%, Thermo Scientific™

Product Code. 15438485
500 mg
Click to view available options
Quantity:
500 mg
This item is not returnable. View return policy

Product Code. 15438485

Brand: Thermo Scientific Alfa Aesar H33464.ME

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 307512-33-0
Melting Point 238°C to 244°C
Molecular Formula C8H8N2S2
MDL Number MFCD01917848
Quantity 500 mg
Synonym 5,6-dimethylthieno 2,3-d pyrimidine-4-thiol, 5,6-dimethyl-thieno 2,3-d pyrimidine-4-thiol, 5,6-dimethylthieno 2,3-d pyrimidine-4 3h-thione, 5,6-dimethyl-3h-thieno 2,3-d pyrimidine-4-thione, thieno 2,3-d pyrimidine-4 3h-thione,5,6-dimethyl, 5,6-dimethylthiopheno 2,3-d pyrimidine-4-thiol, 5,6-dimethyl-thieno 2,3-d pyrimidine-4 3h-thione, 5,6-dimethylthieno 2,3-d pyrimidin-4-ylhydrosulfide, 5,6-dimethyl-3h,4h-thieno 2,3-d pyrimidine-4-thione, hydrosulfide, 5,6-dimethylthieno 2,3-d pyrimidin-4-yl
InChI Key BPXRXYWYEMPYQU-UHFFFAOYSA-N
SMILES CC1=C(SC2=C1C(=S)NC=N2)C
IUPAC Name 5,6-dimethyl-3H-thieno[2,3-d]pyrimidine-4-thione
Molecular Weight (g/mol) 196.286
PubChem CID 727654
Formula Weight 196.29
Percent Purity 96%
Chemical Name or Material 5,6-Dimethylthieno[2,3-d]pyrimidine-4(3H)-thione
Show More Show Less
compliance-icons
Product Identifier
  • 5,6-Dimethylthieno[2,3-d]pyrimidine-4(3H)-thione
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 5,6-Dimethylthieno[2,3-d]pyrimidine-4(3H)-thione
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5,6-Dimethylthieno[2,3-d]pyrimidine-4(3H)-thione, 96%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.