missing translation for 'onlineSavingsMsg'
Learn More
5-Amino-2-methylindole, 97%, Thermo Scientific Chemicals Product Code.: 11400287

5-Amino-2-methylindole, 97%, Thermo Scientific Chemicals

Product Code. 11400287
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11400287

Brand: Thermo Scientific Alfa Aesar B21694.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7570-49-2
Molecular Formula C9H10N2
Molecular Weight (g/mol) 146.19
MDL Number MFCD02093426
InChI Key JQULCCZIXYRBSE-UHFFFAOYSA-N
Synonym 5-amino-2-methylindole, 2-methyl-5-aminoindole, 2-methylindole-5-ylamine, pubchem7273, acmc-209ozq, 2-methylindole-5-amine, 2-methyl-5-amino-1h-indole, bidd:gt0193, 5-amino-2-methyl-1h-indole, 2-methyl-1h-indol-5-ylamineShow MoreShow Less
PubChem CID 2733992
IUPAC Name 2-methyl-1H-indol-5-amine
SMILES CC1=CC2=CC(N)=CC=C2N1

Specifications

Quantity 1 g
Formula Weight 145.19
Percent Purity 97%
Chemical Name or Material 5-Amino-2-methylindole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10129765 Certificate TypeCertificate of Analysis Date27/01/2023 Catalog NumberB21694.03,B21694.14,B21694.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Amino-2-methylindole, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.