missing translation for 'onlineSavingsMsg'
Learn More
5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranoside, 98%, Thermo Scientific Chemicals Product Code.: 10081444

5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranoside, 98%, Thermo Scientific Chemicals

Product Code. 10081444
25 mg, Glass bottle
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10081444

Brand: Thermo Scientific Acros 398920250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2627-69-2
Molecular Formula C9H14N4O5
Molecular Weight (g/mol) 258.23
MDL Number MFCD00869751
InChI Key RTRQQBHATOEIAF-UUOKFMHZSA-N
Synonym acadesine, aica-riboside, aicar, arasine, aica riboside, aic-riboside, acadesina, acadesinum, 5-amino-1-beta-d-ribofuranosyl-1h-imidazole-4-carboxamide, protaraShow MoreShow Less
PubChem CID 17513
ChEBI CHEBI:28498
IUPAC Name 5-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]imidazole-4-carboxamide
SMILES NC(=O)C1=C(N)N(C=N1)[C@@H]1O[C@H](CO)[C@@H](O)[C@H]1O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 214°C to 216°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 mg
Merck Index 15, 17
Specific Rotation -62.0° to -66.0° (20°C, 589nm) (c=1 H2O)
Solubility Information Solubility in water: soluble (7g/L at 25°C). Other solubilities: solubility in aq. pbs (2.5g/L at 25°C),soluble in ethanol,dmso
Formula Weight 258.23
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranoside
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number340235 Certificate TypeCertificate of Analysis Date10/02/2023 Catalog Number398920250,398921000,398920050
Lot Number338838 Certificate TypeCertificate of Analysis Date14/02/2022 Catalog Number398920250,398921000,398920050
compliance-icons
Product Identifier
  • 5-Aminoimidazole-4-carboxamide-1-beta-D-ribofuranoside
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Aminoimidazole-4-carboxamide-1-β-D-ribofuranoside, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.