missing translation for 'onlineSavingsMsg'
Learn More
5-Bromo-2-iodopyrimidine, 97%, Thermo Scientific Chemicals Product Code.: 10189839

5-Bromo-2-iodopyrimidine, 97%, Thermo Scientific Chemicals

Product Code. 10189839
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 10189839

Brand: Thermo Scientific Acros 434250010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 183438-24-6
CAS Min % 96.0
CAS Max % 100.0
Melting Point 100°C to 102°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Molecular Formula C4H2BrIN2
MDL Number MFCD01318111
Quantity 1 g
Synonym 5-bromo-2-iodo-pyrimidine, 2-iodo-5-bromopyrimidine, 5-bromo-2-iodo pyrimidine, pyrimidine, 5-bromo-2-iodo, 5-bromo-2-iodpyrimidine, 5-bromo-2-iodo-1,3-diazine, pubchem6900, 5-brmo-2-iodopyrimidine, acmc-1c1ed, 2-iodo-5-bromo-pyrimidine
InChI Key ZEZKXPQIDURFKA-UHFFFAOYSA-N
SMILES C1=C(C=NC(=N1)I)Br
IUPAC Name 5-bromo-2-iodopyrimidine
Molecular Weight (g/mol) 284.88
PubChem CID 7006651
Formula Weight 284.88
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-iodopyrimidine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number340244 Certificate TypeCertificate of Analysis Date09/08/2021 Catalog Number434250010
compliance-icons
Product Identifier
  • 5-Bromo-2-iodopyrimidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Bromo-2-iodopyrimidine, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.