missing translation for 'onlineSavingsMsg'
Learn More
5-Bromo-2-iodopyrimidine, 98%, Thermo Scientific Chemicals Product Code.: 11358426

5-Bromo-2-iodopyrimidine, 98%, Thermo Scientific Chemicals

Product Code. 11358426
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11358426

Brand: Thermo Scientific Alfa Aesar L17453.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 183438-24-6
Molecular Formula C4H2BrIN2
Molecular Weight (g/mol) 284.882
MDL Number MFCD01318111
InChI Key ZEZKXPQIDURFKA-UHFFFAOYSA-N
Synonym 5-bromo-2-iodo-pyrimidine, 2-iodo-5-bromopyrimidine, 5-bromo-2-iodo pyrimidine, pyrimidine, 5-bromo-2-iodo, 5-bromo-2-iodpyrimidine, 5-bromo-2-iodo-1,3-diazine, pubchem6900, 5-brmo-2-iodopyrimidine, acmc-1c1ed, 2-iodo-5-bromo-pyrimidineShow MoreShow Less
PubChem CID 7006651
IUPAC Name 5-bromo-2-iodopyrimidine
SMILES C1=C(C=NC(=N1)I)Br

Specifications

Melting Point 100°C to 102°C
Quantity 5 g
Sensitivity Light sensitive
Formula Weight 284.88
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-iodopyrimidine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10193588 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL17453.03,L17453.06
Lot NumberI09T005 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL17453.03,L17453.06
Lot NumberW16C001 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL17453.03,L17453.06
Lot Number10236382 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL17453.03,L17453.06
Lot Number10233711 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL17453.03,L17453.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Bromo-2-iodopyrimidine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.