missing translation for 'onlineSavingsMsg'
Learn More
5-Bromoisophthalic acid, 96%, ACROS Organics™ Product Code.: 10277073

5-Bromoisophthalic acid, 96%, ACROS Organics™

Product Code. 10277073
5g; Glass bottle
Click to view available options
5g; Glass bottle
This item is not returnable. View return policy

Product Code. 10277073

Brand: Thermo Scientific Acros 293190050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 23351-91-9
CAS Min % 95.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Molecular Formula C8H3BrO4
Linear Formula BrC6H3(CO2H)2
MDL Number MFCD00152019
Quantity 5 g
Synonym 5-bromoisophthalic acid, 1,3-benzenedicarboxylic acid, 5-bromo, 5-bromoisophthalicacid, pubchem22068, 5-bromo-isophthalic acid, acmc-209g2n, ksc202c0n, 5-bromo-1,3-benzenedicarboxylic acid, 1,3-benzenedicarboxylicacid, 5-bromo
InChI Key JATKASGNRMGFSW-UHFFFAOYSA-L
SMILES [O-]C(=O)C1=CC(=CC(Br)=C1)C([O-])=O
IUPAC Name 5-bromobenzene-1,3-dicarboxylate
Molecular Weight (g/mol) 243.01
PubChem CID 90073
Formula Weight 245.03
Percent Purity ≥95%
Physical Form Powder
Chemical Name or Material 5-Bromoisophthalic acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Bromoisophthalic acid, 96%, ACROS Organics™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.