missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 5 g |
Formula Weight | 228.56 |
Percent Purity | 97% |
Chemical Name or Material | 5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 9
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
5-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylic acid, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction