missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments Repacement Adapter for 16 mm Outer Diameter Tubes

For use with DEN-1 and DEN-1B Densitometer

Brand:  Grant Instruments D16

Product Code. 11451078

  • £44.05 / Each
Estimated Shipment: 26-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • D16 Adaptor
  • For 16mm tubes and the Grant Instruments DEN-1 and DEN-1B Densitometers
.
Specifications

Specifications

DEN-1 and DEN-1B densitometers
16 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Grant Instruments Repacement Adapter for 16 mm Outer Diameter Tubes > For Use With: 16mm O.D. Tubes

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.