missing translation for 'onlineSavingsMsg'
Learn More
5-Fluoro-2-nitrophenylacetic acid, 97%, Thermo Scientific™ Product Code.: 15483397

5-Fluoro-2-nitrophenylacetic acid, 97%, Thermo Scientific™

Product Code. 15483397
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15483397

Brand: Thermo Scientific Alfa Aesar H26126.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29640-98-0
Molecular Formula C8H6FNO4
Molecular Weight (g/mol) 199.14
MDL Number MFCD07787601
InChI Key HOWBVGXZCYNPOU-UHFFFAOYSA-N
Synonym 5-fluoro-2-nitrophenyl acetic acid, 5-fluoro-2-nitrophenylacetic acid, 2-5-fluoro-2-nitrophenyl acetic acid, 5-fluoro-2-nitro-phenyl-acetic acid, 5-fluoro-2-nitrophenylaceticacid, acmc-209z8d, 5-fluoro-2-nitrophenyl aceticacid, 2-nitro-5-fluoro phenylacetic acid, 5-fluoro-2-nitro-phenylacetic acid, 2-nitro-5-fluorophenyl acetic acidShow MoreShow Less
PubChem CID 10397854
IUPAC Name 2-(5-fluoro-2-nitrophenyl)acetic acid
SMILES OC(=O)CC1=C(C=CC(F)=C1)[N+]([O-])=O

Specifications

Melting Point 153°C to 156°C
Quantity 250 mg
Formula Weight 199.14
Percent Purity 97%
Chemical Name or Material 5-Fluoro-2-nitrophenylacetic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10143386 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot Number10140908 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot NumberG15T043 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number10173008 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot NumberF17T020 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Fluoro-2-nitrophenylacetic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.