missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Methyl-1-phenyl-1H-pyrazole-4-sulfonyl chloride, 95+%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
Assay Percent Range | 95% |
Quantity | 1 g |
Formula Weight | 256.71 |
Percent Purity | 95% |
Chemical Name or Material | 5-Methyl-1-phenyl-1H-pyrazole-4-sulfonyl chloride |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
5-Methyl-1-phenyl-1H-pyrazole-4-sulfonyl chloride, 95+%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction