missing translation for 'onlineSavingsMsg'
Learn More
5-Methylisophthalic acid, 98%, Thermo Scientific Chemicals Product Code.: 11497176

5-Methylisophthalic acid, 98%, Thermo Scientific Chemicals

Product Code. 11497176
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11497176

Brand: Thermo Scientific Alfa Aesar B25496.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 499-49-0
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00013986
InChI Key PMZBHPUNQNKBOA-UHFFFAOYSA-N
Synonym 5-methylisophthalic acid, uvitic acid, 5-methyl-1,3-benzenedicarboxylic acid, isophthalic acid, 5-methyl, 1,3-benzenedicarboxylic acid, 5-methyl, uvitinsaure, 3,5-dicarboxytoluene, 5-methylisophthalicacid, acmc-209kj2, ksc237e4tShow MoreShow Less
PubChem CID 68137
IUPAC Name 5-methylbenzene-1,3-dicarboxylic acid
SMILES CC1=CC(=CC(=C1)C(=O)O)C(=O)O

Specifications

Melting Point 296°C to 300°C
Quantity 1 g
Formula Weight 180.16
Percent Purity 98%
Chemical Name or Material 5-Methylisophthalic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252887 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog NumberB25496.03,B25496.06
Lot Number10251745 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberB25496.03,B25496.06
Lot Number10251866 Certificate TypeCertificate of Analysis Date21/08/2024 Catalog NumberB25496.03,B25496.06
Lot Number10235811 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25496.03,B25496.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Methylisophthalic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.