missing translation for 'onlineSavingsMsg'
Learn More
6,7-Dimethoxy-1-methyl-3,4-dihydroisoquinoline, 97%, Maybridge Product Code.: 10557893

6,7-Dimethoxy-1-methyl-3,4-dihydroisoquinoline, 97%, Maybridge

Product Code. 10557893
1g
Click to view available options
1g
This item is not returnable. View return policy

Product Code. 10557893

Brand: Thermo Scientific Maybridge BTB04055DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 4721-98-6
Assay Percent Range 97%
Molecular Formula C12H15NO2
Quantity 1 g
Synonym 1-methyl-6,7-dimethoxy-3,4-dihydroisoquinoline, isoquinoline, 3,4-dihydro-6,7-dimethoxy-1-methyl, 6,7-dimethoxy-1-methyl-3,4-dihydro-isoquinoline, dehydrosalsolidine, maybridge1_001068, 3,4-dihydro-6,7-dimethoxy-1-methylisoquinoline, vasuqtgzapzkfk-uhfffaoysa, 1-methyl-3,4-dihydro-6,7-dimethoxyisoquinoline, 1-methyl-6?7-dimethoxy-3?4-dihydroisoquinoline, #
InChI Key VASUQTGZAPZKFK-UHFFFAOYSA-N
SMILES CC1=NCCC2=CC(=C(C=C12)OC)OC
IUPAC Name 6,7-dimethoxy-1-methyl-3,4-dihydroisoquinoline
Molecular Weight (g/mol) 205.257
PubChem CID 22652
Formula Weight 205.26
Chemical Name or Material 6, 7-Dimethoxy-1-methyl-3, 4-dihydroisoquinoline
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6,7-Dimethoxy-1-methyl-3,4-dihydroisoquinoline, 97%, Maybridge >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.