missing translation for 'onlineSavingsMsg'
Learn More
6-Boc-1-oxa-6-azaspiro[3.3]heptane, 96%, Thermo Scientific Chemicals Product Code.: 15483588

6-Boc-1-oxa-6-azaspiro[3.3]heptane, 96%, Thermo Scientific Chemicals

Product Code. 15483588
250 mg
Click to view available options
Quantity:
250 mg
500 mg
This item is not returnable. View return policy

Product Code. 15483588

Brand: Thermo Scientific Alfa Aesar H57040.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1223573-41-8
Molecular Formula C10H17NO3
Molecular Weight (g/mol) 199.25
MDL Number MFCD16877674
InChI Key QOXFRPSIASGKGE-UHFFFAOYSA-N
Synonym tert-butyl 1-oxa-6-azaspiro 3.3 heptane-6-carboxylate, 1-oxa-6-azaspiro 3.3 heptane-6-carboxylic acid tert-butyl ester, 6-boc-1-oxa-6-azaspiro 3.3 heptane, tert-butyl2,5-dioxa-8-aza-spiro 3,5 nonane-8-carboxylate, 2-aza-5-oxaspiro 3.3 heptane-2-carboxylic acid tert-butyl ester, 1-oxa-6-azaspiro 3.3 heptane-6-carboxylic acid,1,1-dimethylethyl ester, tert-butyl 1-oxa-6-azaspiro 3.3 heptane-6-carboxylate 95+%Show MoreShow Less
PubChem CID 54758968
IUPAC Name tert-butyl 1-oxa-6-azaspiro[3.3]heptane-6-carboxylate
SMILES CC(C)(C)OC(=O)N1CC2(CCO2)C1

Specifications

Quantity 250 mg
Formula Weight 199.25
Percent Purity 96%
Chemical Name or Material 6-Boc-1-oxa-6-azaspiro[3.3]heptane

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberB13Y059 Certificate TypeCertificate of Analysis Date13/01/2025 Catalog NumberH57040.MD
Lot NumberB13Y059 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH57040.MD,H57040.ME

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6-Boc-1-oxa-6-azaspiro[3.3]heptane, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.