missing translation for 'onlineSavingsMsg'
Learn More
6-Chloro-2-benzoxazolethiol, 99%, Thermo Scientific Chemicals Product Code.: 10581521

6-Chloro-2-benzoxazolethiol, 99%, Thermo Scientific Chemicals

Product Code. 10581521
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10581521

Brand: Thermo Scientific Acros 297300050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22876-20-6
Molecular Formula C7H4ClNOS
Molecular Weight (g/mol) 185.63
MDL Number MFCD01456482
InChI Key HAASPZUBSZGCKU-UHFFFAOYSA-N
Synonym 6-chloro-1,3-benzoxazole-2-thiol, 6-chloro-2-mercaptobenzoxazole, 6-chloro-2-benzoxazolethiol, 6-chlorobenzo d oxazole-2-thiol, 2-mercapto-6-chlorobenzoxazole, 6-chlorobenzoxazole-2 3h-thione, 6-chlorobenzo d oxazole-2 3h-thione, 6-chloro-benzooxazole-2-thiol, pubchem8714, 6-chlorobenzoxazole-2-thiolShow MoreShow Less
PubChem CID 2119214
IUPAC Name 6-chloro-3H-1,3-benzoxazole-2-thione
SMILES C1=CC2=C(C=C1Cl)OC(=S)N2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 229.0°C to 232.0°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: .. Other solubilities: soluble in solutions of bases,soluble in ethanol
Formula Weight 185.63
Percent Purity 99%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 6-Chloro-2-benzoxazolethiol, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0190955 Certificate TypeCertificate of Analysis Date24/10/2022 Catalog Number297300050,297300250

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
6-Chloro-2-benzoxazolethiol, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.