Learn More
Ansell Edmont™ 76-501 Series Blue FiberTuf™ Light Weight Gloves Product Code.: 12696991
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
Ansell Edmont™ 76-501 Series Blue FiberTuf™ Light Weight Gloves
Product Code. 12696991 Shop All Ansell Products
10
Click to view available options
Size:
7
8
9
10
Product Code. 12696991
Brand: Ansell Edmont™ 76501100
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
Add to basket
Designed for optimal results in dry applications requiring light mechanical protection, comfort and good grip.
  • Aromatic polyester fiber construction
  • For handling, packaging, light assembly applications
  • Synthetic fiber liner
  • Knitted construction
  • PVC coating
  • Dotted on palm side finish
  • Knitwrist cuff
TRUSTED_SUSTAINABILITY
Specifications
Back Material PVC
Certifications/Compliance Standard CATEGORY II EN 388 : 214X
Cuff Material PVC
Color Blue
Product Type Lightweight Gloves
Size 10
Glove Cuff Style Knit
Lining Polyester Cotton
Palm Type Dotted-palm
Quantity 12 Pairs
Series 76-501
Show More Show Less
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131
    Expires2025-06-08
    New Offer
    Shop Now
Product Content Correction
Product Title
Ansell Edmont™ 76-501 Series Blue FiberTuf™ Light Weight Gloves >