Learn More
Thermo Scientific™ 8mm Autosampler Inserts
Clear glass or polypropylene inserts for 8mm HPLC and GC autosampler vials are available in a variety of configurations with usable volumes from 125μL to 200μL.
£32.55 - £364.00
Specifications
Color | Clear |
---|---|
Certifications/Compliance | Certificate of Analysis (COA), Certificate of Conformance (COC) |
Product Code | Brand | Volume (Metric) | Material | Base Style | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Material | Base Style | Quantity | Price | Quantity & Availability | ||||||
10119084
|
Thermo Scientific™
C4012-465 |
250 μL | Glass | Flat Bottom | 500 | N/A | N/A | N/A | |||||
10538215
|
Thermo Scientific™
C4012-530 |
200 μL | Silanized Glass | Conical | 100 | N/A | N/A | N/A | |||||
10330571
|
Thermo Scientific™
401BS-530 |
200 μL | Silanized Glass | Conical | 100 |
£73.10
Pack of 100 |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||||
12470088
|
Thermo Scientific™
C4012-530P |
175 μL | Polypropylene | Polyspring | 100 | N/A | N/A | N/A | |||||
12440088
|
Thermo Scientific™
C4012-529 |
200 μL | Glass | Conical | 100 |
£34.48
Pack of 100 |
In Stock
Log in to see stock. ![]() |
||||||
12450098
|
Thermo Scientific™
C4012-S530 |
200 μL | Silanized Glass | Conical | 100 | N/A | N/A | N/A | |||||
10185482
|
Thermo Scientific™
MTS-1 |
125 to 200 μL | Polyethylene | 500 | N/A | N/A | N/A | ||||||
10227672
|
Thermo Scientific™
02-NV |
250 μL | Glass | Flat Bottom | 1000 | N/A | N/A | N/A | |||||
10728544
|
Thermo Scientific™
02-MTV |
200 μL | Glass | Conical | 1000 | N/A | N/A | N/A | |||||
Description
These clear glass or polypropylene inserts for 8mm HPLC and GC autosampler vials are available in a variety of configurations with usable volumes ranging from 125-200μL.
- Pulled point inserts are an economical choice for non-critical applications
Specifications
Clear | |
Certificate of Analysis (COA), Certificate of Conformance (COC) |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.