missing translation for 'onlineSavingsMsg'
Learn More
9-Fluorenone, 99+%, Thermo Scientific Chemicals Product Code.: 10754041

9-Fluorenone, 99+%, Thermo Scientific Chemicals

Product Code. 10754041
500 g, Plastic bottle
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10754041

Brand: Thermo Scientific Acros 119205000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 486-25-9
InChI Key YLQWCDOCJODRMT-UHFFFAOYSA-N
Synonym 9-fluorenone, 9h-fluoren-9-one, fluorenone, 9-oxofluorene, diphenylene ketone, unii-az9t83s2aq, ccris 593, 9h-fluorene-9-one, az9t83s2aq, 9-fluoreneoneShow MoreShow Less
PubChem CID 10241
ChEBI CHEBI:17922
IUPAC Name fluoren-9-one
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C2=O

Specifications

Melting Point 81.0°C to 85.0°C
Color Yellow
Boiling Point 342.0°C
Flash Point 163°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Molecular Formula C13H8O
Quantity 500 g
Beilstein 07,465
Solubility Information Solubility in water: insoluble.
Molecular Weight (g/mol) 180.21
Formula Weight 180.21
Percent Purity 99+%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 9-Fluorenone
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0430323 Certificate TypeCertificate of Analysis Date20/11/2023 Catalog Number119200050,119201000,119205000,119200025
Lot NumberA0398320 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number119200050,119201000,119205000,119200025
Lot NumberA0409468 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number119200050,119201000,119205000,119200025
Lot NumberA0407869 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number119200050,119201000,119205000,119200025
Lot NumberA0413370 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number119200050,119201000,119205000,119200025

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
9-Fluorenone, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.