Learn More
BINDER™ Temperature Calibration Certificate with 9 Measurement Points
For Use With: BINDER units series All ED, FD, FED, FP, M, FDL, MK, MKT, BD, BF, KB, C, CB, KBW
Brand:  BINDER™ DL30-0109
Product Code. 15642306
  • £1160.00 / Each
Explore more special offers
Description
Description
  • Measurement at specified temperature
  • Including certificate
  • Various points of measurement
Specifications
Specifications
BINDER Chambers Series All ED, FD, FED, FP, M, FDL, MK, MKT, BD, BF, KB, C, CB, KBW
Temperature measurement, 9 measuring points - Temperature measurement with 9 measuring points with a set value specified by the user, including certificate
Temperature Calibration Certificate
Temperature Measurement
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BINDER™ Temperature Calibration Certificate with 9 Measurement Points > 9 measurement points; For Use With: All ED, FD, FED, FP, M, FDL, MDL, MK, MKT, BD, BF, KB, BFD, BFED, C, CB, KBW models