Learn More
BINDER™ Access Port
Access port with silicone plug for use with KB PRO 130, KB PRO 260, KB PRO 720, KB PRO 1060, and KB PRO 1600.
Brand: BINDER™ 8012-2478
This item is not returnable.
View return policy
Description
- Round access port with silicone plug
- Choose between 30, 50, or 100 mm plug
- Available on left, right, or top of the chamber
- For use with KB PRO 130, KB PRO 260, KB PRO 720, KB PRO 1060, and KB PRO 1600
Specifications
KB PRO 130, KB PRO 260, KB PRO 720, KB PRO 1060, KB PRO 1600 | |
Access Port | |
100 mm |
Silicone | |
1 on Right Side |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
BINDER™ Access Port > Access Port, 100 mm, Right
Spot an opportunity for improvement?Share a Content Correction