Learn More
BINDER™ Access Port
Access port with silicone plug for use with KBF PRO 130, KBF PRO 260, KBF PRO 470, KBF PRO 720, KBF PRO 1060, and KBF PRO 1600.
Brand: BINDER™ 8012-2495
Description
- Round access port with silicone plug
- Choose between 30, 50, or 100 mm plug
- Available on left, right, or top of the chamber
- For use with KBF PRO 130, KBF PRO 260, KBF PRO 470, KBF PRO 720, KBF PRO 1060, and KBF PRO 1600
Specifications
KBF PRO 130, KBF PRO 260, KBF PRO 470, KBF PRO 720, KBF PRO 1060, KBF PRO 1600 | |
Access Port | |
50 mm |
Silicone | |
1 on Top |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.