missing translation for 'onlineSavingsMsg'
Learn More
Acetonitrile, Optima™ LC/MS Grade, Fisher Chemical™ Product Code.: 10055454

Acetonitrile, Optima™ LC/MS Grade, Fisher Chemical™

Product Code. 10055454
500 mL
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
This item is not returnable. View return policy

Product Code. 10055454

Brand: Fisher Chemical A955500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Acetonitrile, >-99.9%, C2H3N, CAS Number-75-05-8, methyl cyanide, cyanomethane, acetonitrile, ethyl nitrile, ethanenitrile, cyanure de methyl, methanecarbonitrile, methylkyanid, methane, cyano, MeCN

Acetonitrile, >-99.9%, C2H3N, CAS Number-75-05-8, methane, cyano, ethyl nitrile, methanecarbonitrile, methylkyanid, mecn, acetonitrile, methyl cyanide, ethanenitrile, cyanure de methyl, cyanomethane

  • Ideal mobile phase for LC/MS and UHPLC-UV applications in pharmaceutical, biotechnology, clinical, environmental and food safety industries.
  • Novel gradient elution test with Photodiode Array Detector screens for UV-absorbing contaminants at every wavelength in the 200 to 400nm range to afford smooth baselines and to reduce interferences.
  • Evaluated for 17 metal impurities at ppb concentrations to minimize the formation of metal adducts during analysis and simplify spectral interpretation.
  • 0.1 micron filtration minimizes troublesome particles that may interfere with LC/MS and UHPLC system performance
  • Provides ideal solution to prepare cleaner, particle-free mobile phase blends. Positive mode ionization (as propazine) and negative mode ionization (as mecoprop) with ESI and Single Quadrupole MS detect organic contaminants at max 50ppb and max 300ppb, respectively
    • Glass bottles have fluorinated lining
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Weight (g/mol) 41.053
MDL Number 1878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecnShow MoreShow Less
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -46°C
Absorbance ≤1.00 AU at 190nm, ≤0.15 AU at 195nm, ≤0.05 AU at 200nm, ≤0.04 AU at 205nm, ≤0.03 AU at 210nm, ≤0.025 AU at 215nm, ≤0.015 AU at 220nm, ≤0.015 AU at 225nm, ≤0.01 AU at 230nm, ≤0.005 AU at 254nm, ≤0.005 AU at 280nm
Color Colorless
Decomposition Information >500°C
Density 0.781g/cm³
Boiling Point 81°C
Quantity 500 mL
Identification Pass Test
Packaging Glass Bottle with fluorinated lining
Physical Form Liquid
Assay Percent Range Assay: 99.9% min by GC
Residue after Evaporation ≤0.8ppm
Molecular Formula C2H3N
Titratable Acid ≤0.008meq/g
Linear Formula CH3CN
Titratable Base ≤0.0006meq/g
Vapor Pressure 97mbar at 20°C
UN Number 1648
Viscosity 0.36cp at 20°C
Water ≤0.01%
Merck Index 15, 69
Formula Weight 41.05
Percent Purity ≥99.9%
Grade Formulated for UHPLC-UV
Assay GC
Chemical Name or Material Acetonitrile
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2528083 Certificate TypeCertificate of Analysis Date26/02/2025 Catalog NumberA955-500
Lot Number2528358 Certificate TypeCertificate of Analysis Date25/02/2025 Catalog NumberA955-500
Lot Number2528091 Certificate TypeCertificate of Analysis Date24/02/2025 Catalog NumberA955-500
Lot Number2528090 Certificate TypeCertificate of Analysis Date24/02/2025 Catalog NumberA955-500
Lot Number2527293 Certificate TypeCertificate of Analysis Date18/02/2025 Catalog NumberA955-500
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Acute toxicity Category 4
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetonitrile, Optima™ LC/MS Grade, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.