missing translation for 'onlineSavingsMsg'
Learn More
Acetophenone, 99%, Thermo Scientific Chemicals Product Code.: 15463057

Acetophenone, 99%, Thermo Scientific Chemicals

Product Code. 15463057
500 mL
Click to view available options
Quantity:
10,000 mL
2500 mL
500 mL
This item is not returnable. View return policy

Product Code. 15463057

Brand: Thermo Scientific Alfa Aesar A12727.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Acetophenone is used as a raw material in the synthesis of active pharmaceutical ingredients such as dextropropoxyphene and phenylpropanolamine. It is employed in organic synthesis and as a polymerization catalyst for olefins. It is involved in the preparation of styrene, 1-phenylethanol. It is also used as a active component of coatings and inks. It reacts with formaldehyde to prepare resins. Further, it is used in textile coatings, in perfumes for orange blossom odor. In addition to this, it is used as a solvent in plastics, adhesives and a flavoring agent in foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetophenone is used as a raw material in the synthesis of active pharmaceutical ingredients such as dextropropoxyphene and phenylpropanolamine. It is employed in organic synthesis and as a polymerization catalyst for olefins. It is involved in the preparation of styrene, 1-phenylethanol. It is also used as a active component of coatings and inks. It reacts with formaldehyde to prepare resins. Further, it is used in textile coatings, in perfumes for orange blossom odor. In addition to this, it is used as a solvent in plastics, adhesives and a flavoring agent in foods.

Solubility
Miscible with water, chloroform, ethanol, ether, benzene and acetone. Slightly miscible with concentrated sulfuric acid.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-86-2
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.151
MDL Number MFCD00008724
InChI Key KWOLFJPFCHCOCG-UHFFFAOYSA-N
Synonym acetophenone, methyl phenyl ketone, acetylbenzene, phenyl methyl ketone, ethanone, 1-phenyl, hypnone, benzoyl methide, acetophenon, 1-phenylethan-1-one, acetylbenzolShow MoreShow Less
PubChem CID 7410
ChEBI CHEBI:27632
IUPAC Name 1-phenylethanone
SMILES CC(=O)C1=CC=CC=C1

Specifications

Melting Point 19°C to 20°C
Density 1.03
Boiling Point 201°C to 203°C
Flash Point 76°C (169°F)
Linear Formula C6H5COCH3
Refractive Index 1.5325
Quantity 500 mL
Merck Index 14,73
Solubility Information Miscible with water,chloroform,ethanol,ether,benzene and acetone. Slightly miscible with concentrated sulfuric acid.
Formula Weight 120.15
Percent Purity 99%
Chemical Name or Material Acetophenone
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252186 Certificate TypeCertificate of Analysis Date02/09/2024 Catalog NumberA12727.AP,A12727.0F,A12727.0D
Lot Number10241090 Certificate TypeCertificate of Analysis Date12/04/2024 Catalog NumberA12727.AP,A12727.0F,A12727.0D
Lot Number10181102 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12727.AP,A12727.0F,A12727.0D
Lot Number10196775 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12727.AP,A12727.0F,A12727.0D
Lot NumberD7422B Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12727.AP,A12727.0F,A12727.0D
compliance-icons
Product Identifier
  • Acetophenone, 99%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Acetophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acetophenone, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.