missing translation for 'onlineSavingsMsg'
Learn More
Acrylic acid, 98%, extra pure, stabilized, Thermo Scientific Chemicals Product Code.: 10163890

Acrylic acid, 98%, extra pure, stabilized, Thermo Scientific Chemicals

Product Code. 10163890
1 L, Glass bottle
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10163890

Brand: Thermo Scientific 164250010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-10-7, 150-76-5
Molecular Formula C3H4O2
Molecular Weight (g/mol) 72.06
MDL Number MFCD00004367
InChI Key NIXOWILDQLNWCW-UHFFFAOYSA-N
Synonym acrylic acid, 2-propenoic acid, propenoic acid, vinylformic acid, acroleic acid, propene acid, ethylenecarboxylic acid, polyacrylate, propenoate, carbomerShow MoreShow Less
PubChem CID 6581
ChEBI CHEBI:18308
IUPAC Name prop-2-enoic acid
SMILES C=CC(=O)O

Specifications

CAS Min % 98
Melting Point 13.0°C
Density 1.0500g/mL
Boiling Point 139.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCO2H
Refractive Index 1.4192 to 1.4212
Quantity 1 L
Beilstein 02, 397
Merck Index 15, 121
Specific Gravity 1.05
Solubility Information Solubility in water: miscible. Other solubilities: miscible with most organic solvents,soluble in acetone
Stabilizer 180 to 220ppm MEHQ
Viscosity 1.3 mPa.s (20°C)
Formula Weight 72.06
Percent Purity 98%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Acrylic acid, Stabilized
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464678 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number164250010,164250100,164252500,164250025
Lot NumberA0464678 Certificate TypeCertificate of Analysis Date26/07/2024 Catalog Number164250010,164250100,164252500,164250025
Lot NumberA0459748 Certificate TypeCertificate of Analysis Date21/02/2024 Catalog Number164250010,164250100,164252500,164250025
Lot NumberA0452743 Certificate TypeCertificate of Analysis Date03/11/2023 Catalog Number164250010,164250100,164252500,164250025
Lot NumberA0452744 Certificate TypeCertificate of Analysis Date23/10/2023 Catalog Number164250010,164250100,164252500,164250025
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Acrylic acid, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Acrylic acid, 98%, extra pure, stabilized, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.