missing translation for 'onlineSavingsMsg'
Learn More

Socorex™ Acura™ Manual XS Micropipettes

Superior, extra sharp micropipettes that are short, smooth and lightweight. Socorex™ Acura™ Manual XS Micropipettes are ergonomically designed and optimally suited for research laboratories.

Brand:  Socorex™ 826.0010

 View more versions of this product

Product Code. 11972431

  • £302.00 / Each
Estimated Shipment: 22-07-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Optimized shape/size ratio for ease of use when working with micro tubes.
  • Reduced weight - lighter than leading brands.
  • Unique, Socorex™ patented lip seal reduces activation forces and provides for ultra-soft pipetting.
  • Adjustable tip ejector.
  • Autoclavable fully assembled.
  • Unsurpassed metrologic performance.
Specifications

Specifications

<±2.5% - <±1.0%
XS Manual
Acura XS
0.5 to 10 μL
1 Each
Autoclavable
1
Micropipette
<1.8% - <0.5%
36 Months
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Socorex™ Acura™ Manual XS Micropipettes >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.