missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Adhesive Mats for Solaris™ Open Air Shakers & Temperature Controlled Shakers, and MaxQ™ 6000 Shakers

Mix and match Thermo Scientific™ Solaris™ shaker Universal clamps to suit individual needs in the laboratory. Adhesive mats are compatible with all Thermo Scientific™ Solaris and MaxQ platforms and can be cut to size

£45.09 - £67.61


Products 5
Product Code Brand Description Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Description Dimensions (L x W) Price Quantity & Availability  
17811895
View Documents Promotion Details
Thermo Scientific™
75004111
"2 Low" Adhesion Mat (one single mat) 200 x 200 mm
£45.09
Each
Estimated Shipment: 11-04-2025
to see stock.
Add to basket
 
17811905
View Documents
Thermo Scientific™
75004117
"2 Low" Adhesion Mat (one single mat) 280 x 170 mm
£59.42
Each
Estimated Shipment: 11-04-2025
to see stock.
Add to basket
 
17881885
View Documents Promotion Details
Thermo Scientific™
75004126
"2 High" Adhesion Mat (one single mat) 200 x 200 mm
£49.45
Each
Estimated Shipment: 14-04-2025
to see stock.
Add to basket
 
17861905
View Documents Promotion Details
Thermo Scientific™
75004127
"2 High" Adhesion Mat (one single mat) 280 x 170 mm
£67.61
Each
Estimated Shipment: 11-04-2025
to see stock.
Add to basket
 
11706449
View Documents Promotion Details
Thermo Scientific™
300349
Adhesive Flask Mat 23 x 23 cm (9 x 9 in.)
£50.66
Each
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

Mix and match Thermo Scientific™ Solaris™ shaker accessories to suit individual needs in the laboratory. Adhesive mats are compatible with all Thermo Scientific™ Solaris and MaxQ platforms and can be cut to size.

Mix and match Thermo Scientific™ Solaris™ shaker accessories to suit individual needs in the laboratory.  Adhesive mats are compatible with all Thermo Scientific™ Solaris and MaxQ platforms.  

  • Adhesive mats can be cut to any desired size
  • Suitable for use with containers with a wide, flat base, such as flasks, media bottles, microplates, petri dishes, and beakers
  • Low adhesion mats are transparent and are suitable for all vessels up to a maximum of 200 rpm*
  • High adhesion mats are black opaque and are suitable for all vessels up to a maximum of 400 rpm*
  • The rolling applicator tool is designed to help press down the adhesive mats to the platform

* The maximum speed depends on the type of vessel, as distinguished by size, shape, bottom shape, material, surface roughness and cleanliness. You must evaluate these parameters  to  determine  the  optimum  choice  of  mat  for  your  application  before  using the shaker. Additional guidelines on cleaning, installation, and use can be found in the Thermo Scientific™ Solaris™ Shaker Series Instruction Manual

Videos
SDS
Documentos

Documentos

Product Certifications
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Thermo Scientific™ Adhesive Mats for Solaris™ Open Air Shakers & Temperature Controlled Shakers, and MaxQ™ 6000 Shakers

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado