Learn More
METTLER TOLEDO Advanced MR Analytical Balance
METTLER TOLEDO Advanced MR Analytical Balances offer a highly precise weigh cell with FACT automatic internal adjustment.
Brand:  METTLER TOLEDO 30666203
 View more versions of this product
Product Code. 18042294
  • £3910.00 / Each
Explore more special offers
Description
Description
User management within the 4.5-inch touchscreen increases efficiency of daily tasks with 8 built-in applications and statistical data analysis. Designed with full metal housing and overload protection, these balances will provide high quality performance for years to come. When your balance is not in use, enable the power-saving mode to reduce energy consumption and save costs.
MR Analytical Balances include:
  • FACT automatic internal adjustment
  • 4.5 inch color touchscreen
  • 8 built-in applications
  • Statistical data analysis
  • User management
  • Guided leveling
  • 4 interfaces: USB-A, USB-C, RS232, Ethernet
  • Bluetooth option
  • Full metal housing
  • Overload protection
The MR Balance is the successor to the ML or ME-T Balance. See how it ranks among our balance lines:
  • XPR Excellence Analytical Balances
  • MX Advanced Analytical Balances
  • MR Advanced Analytical Balances
  • MA Standard Analytical Balances
  • ME Standard Analytical Balances
  • LA Standard Analytical Balances
TRUSTED_SUSTAINABILITY
Specifications
Specifications
USB-A, USB-C, RS232, Ethernet
Color Touchscreen
Metal
2 sec.
100/240 V, 50/60 Hz
120 g
0.1 mg
MR104
Analytical Balance
No
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
METTLER TOLEDO Advanced MR Analytical Balance > Model: MR104; Capacity: 120 g; Readability: 0.1 mg