Learn More
AGO2 Mouse anti-Human, Unconjugated, Clone: 1G2H12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67934-1-IG-150UL
Description
The AGO2 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human AGO2. This antibody recognizes human, mouse, rat antigen. The AGO2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
AGO2 | |
Monoclonal | |
Unconjugated | |
150 μL | |
27161 | |
IgG1 |
Western Blot, Immunohistochemistry | |
1G2H12 | |
Mouse | |
Primary | |
Human, Mouse, Rat |
Product Suggestions
Customers who viewed this item also viewed