Learn More
Asecos™ Air Purifier PURIFIAIR.488
Air Purifier PURIFIAIR.488 reduces indoor pollutant levels with a multi-component filter system.
Brand: Asecos™ 39323
Additional Details : Weight : 1.40000kg
Description
- Asecos PURIFIAIR.488 can be used to reduce pollutant levels in indoor atmospheres
- Removes mould spores, pollen, and allergens
- Eliminates germs and bacteria and neutralises them
- Continuously filters out fine dust particles
- Removes unpleasant odours and harmful gases from indoor atmospheres
- Built-in PM2.5 sensor and colour display allow users to quickly identify and easily read off the particle concentration in the indoor atmosphere in four levels from green (low) to red (high)
- 1 device handles a volume flow rate of up to 488 m3/h and is designed for approx. 80 m3 room volume (corresponding to a surface area of 32 m2; at a room height of 2.5 m)
- A larger number of devices should be used in larger rooms
- Fresh air must be supplied to the rooms via natural or technical ventilation
- Air purifiers should not be considered a substitute for a fresh air supply
- Efficient results over time only if the filter is replaced at regularly according to the electronic display
- Large filter units ensure a long service life
- Low energy requirements result in low usage costs
- Integrated humidification for a healthy and comfortable indoor climate
Specifications
Prefilter, Cold Catalysis Filter, Activated Carbon Filter, High-performance Particle Filter | |
1.519 Kg | |
559 mm | |
Replacement Filter |
AIRPURIFIAIR.488 | |
83 mm | |
426 mm |