missing translation for 'onlineSavingsMsg'
Learn More

Syngene™ Airclean 600 PCR Tall Workstation
GREENER_CHOICE

Airclean 600 PCR Tall Workstation with UVTect microprocessor controller and vertical laminar air flow.

Brand:  Syngene™ SYN-32PWTLF-E

5715.78 GBP valid until 2025-04-30
Use promo code "24119" to get your promotional price.


 View more versions of this product

Product Code. 16524341

  • £6735.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Tall version
  • With UVTect microprocessor controller
  • Provides Class 100 vertical laminar air flow and timed uv light for sterilization
  • Includes HEPA filter, initial prefilter, fluorescent light, UV shelf with pipette holder, safety switch, and slip hatch
Specifications

Specifications

Tall Workstation
UV Controller
32 in.
UL, CSA, and CE approved
220 V
81.28 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Syngene™ Airclean 600 PCR Tall Workstation > Width: 32 in. (81.28 cm)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.