Learn More
3M™ Petrifilm™ Rapid E.coli / Coliform Count Plate
Povide a cost-effective, convenient and reliable method for testing equipment, raw materials, food products and manufacturing environmental samples.
Brand: 3M™ Petrifilm™ 7100127434
This item is not returnable.
View return policy
Description
- E. coli and coliform results in 18-24 hours
- Plates are sample-ready, eliminating the time-consuming, cumbersome step of preparing media/agar dishes.
- Can be used with the 3M™ Petrifilm™ Plate Reader for consistent, fast, automated reading and recording of results.
- This plate has been awarded Official Methods of Analysis (OMA) #2018.13 for select food and MicroVal certificate no. 2017LR76 for a broad range of foods and select environmental surfaces by Lloyd's Register Nederland B.V. following ISO 16140-2. Food Categories tested: meat products, dairy and egg products, fish and seafood products, vegetable products, ready to eat and ready to reheat, animal feed and samples from the industrial production environment
- Food processors and Contract labs worldwide trust 3M Petrifilm Plates to provide accurate, efficient, consistent results.
Specifications
Count Plate | |
Environmental Test | |
50 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
3M™ Petrifilm™ Rapid E.coli / Coliform Count Plate
Spot an opportunity for improvement?Share a Content Correction