missing translation for 'onlineSavingsMsg'
Learn More
Albendazole, 98+%, Thermo Scientific Chemicals Product Code.: 11415680

Albendazole, 98+%, Thermo Scientific Chemicals

Product Code. 11415680
10 g
Click to view available options
Quantity:
10 g
50 g
100 g
This item is not returnable. View return policy

Product Code. 11415680

Brand: Thermo Scientific Alfa Aesar H25925.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54965-21-8
Molecular Formula C12H15N3O2S
Molecular Weight (g/mol) 265.33
MDL Number MFCD00083232
InChI Key HXHWSAZORRCQMX-UHFFFAOYSA-N
Synonym albendazole, albenza, eskazole, valbazen, zentel, albendazol, proftril, albendazolum, bilutac, zental
PubChem CID 2082
ChEBI CHEBI:16664
IUPAC Name methyl N-(6-propylsulfanyl-1H-benzimidazol-2-yl)carbamate
SMILES CCCSC1=CC=C2N=C(NC(=O)OC)NC2=C1

Specifications

Melting Point 208°C to 210°C
Quantity 10 g
Merck Index 14,210
Formula Weight 265.34
Percent Purity ≥98%
Chemical Name or Material Albendazole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM16L011 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberH25925.18,H25925.09,H25925.22
Lot NumberY11K015 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog NumberH25925.18,H25925.09,H25925.22
Lot Number10252692 Certificate TypeCertificate of Analysis Date07/11/2024 Catalog NumberH25925.18,H25925.09,H25925.22
Lot NumberU12K016 Certificate TypeCertificate of Analysis Date28/08/2024 Catalog NumberH25925.18,H25925.09,H25925.22
Lot NumberQ30K023 Certificate TypeCertificate of Analysis Date07/05/2024 Catalog NumberH25925.18,H25925.09,H25925.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Albendazole, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.