Learn More
Alconox™ Liquinox™ Critical Cleaning Liquid Detergent
Concentrated, mild, anionic detergent for manual and ultrasonic cleaning
Brand: Alconox™ 17775-40
Description
This concentrated, mild, anionic detergent is suitable for manual and ultrasonic cleaning.
Liquinox consists primarily of a homogeneous blend of sodium linear alkylaryl sulfonate, sodium xylene sulfonate, alkanolamide, and ethoxylated alcohol.
- Concentrated
- Phosphate free, biodegradable and readily disposable
- Completely soluble in hard and soft water
- Replaces corrosive acids and hazardous solvents
- Dilute 1:100 (dilute 10mL in 1L of water)
- Free rinsing—gives reliable results without interfering residues
- Use to pass your cleaning validation tests for lab accreditation and plant inspection approval
- USDA authorized for use in federally inspected meat and poultry plants
Used to clean:
Healthcare instruments, laboratory ware, vacuum equipment, tissue culture ware, personal protective equipment, sampling apparatus, catheters, tubing, wine glasses, clean rooms, medical devices, optical parts, electronic components, pharmaceutical apparatus, cosmetics manufacturing equipment, metal castings, forgings and stampings, industrial parts, pipes, tanks and reactors. Passes inhibitory residue test for water analysis. Used for phosphate sensitive analysis ware.
Used to remove:
Soil, grit, grime, slime, grease, oils, blood, tissue, particulates, deposits, chemicals and solvents.
Surfaces cleaned:
Corrosion inhibited formulation recommended for glass, metal, stainless steel, porcelain, ceramic, plastic, cement and fiberglass. Can be used on soft metals such as copper, aluminum, zinc and magnesium if rinsed promptly. Used for art restoration.
Cleaning method:
Soak, brush, sponge, cloth, ultrasonic, flow through clean-inplace. Will foam—not for spray or machine use.
Shelf life: 2 years from date of manufacture
Order Info
3.8L (1 gal.) makes 378.5L (100 gal.) cleaning solution.
Specifications
Critical Cleaning Liquid Detergent | |
8.5 | |
Anionic Detergent, Sodium Linear Alkylaryl Sulfonate, Sodium Xylene Sulfonate, Alkanolamide, Ethoxylated Alcohol, Water | |
Store closed and upright in a cool dry place at 15° to 30°C. | |
1 gal. |
Liquid | |
Pale Yellow | |
Case | |
Clean healthcare instruments, laboratory ware, vacuum equipment, tissue culture ware | |
3.78 L |
Safety and Handling
- Liquinox
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains :
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70% -
Multiple Manufacturers
Chemicals Monthly Promotion
Take advantage of our exclusive monthly promotions on high-quality chemicals.Promo Code: 24199Expires: 2025-04-30
New Offer Save Up to 59%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.