Learn More
Allyl bromide, 99%, stabilized, Thermo Scientific Chemicals
£29.65 - £515.57
Chemical Identifiers
CAS | 106-95-6, 75-56-9 |
---|---|
Molecular Formula | C3H5Br |
Molecular Weight (g/mol) | 120.98 |
MDL Number | MFCD00000244 |
InChI Key | BHELZAPQIKSEDF-UHFFFAOYSA-N |
Synonym | allyl bromide, 3-bromopropene, 1-propene, 3-bromo, 3-bromopropylene, 3-bromo-1-propene, bromallylene, 2-propenyl bromide, 1-bromo-2-propene, propene, 3-bromo, allylbromide |
PubChem CID | 7841 |
IUPAC Name | 3-bromoprop-1-ene |
SMILES | C=CCBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10706131
|
Thermo Scientific Acros
102902500 |
250 mL | Glass bottle |
£29.65
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10448240
|
Thermo Scientific Acros
102905000 |
500 mL | Glass bottle |
£41.41
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10528470
|
Thermo Scientific Acros
102900025 |
2.5 L | Glass bottle |
£148.74
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10538470
|
Thermo Scientific Acros
102900050 |
5 L | Metal Can |
£346.00
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10609283
|
Thermo Scientific Acros
102900100 |
10 L | Plastic drum |
£515.57
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentChemical Identifiers
106-95-6, 75-56-9 | |
120.98 | |
BHELZAPQIKSEDF-UHFFFAOYSA-N | |
7841 | |
C=CCBr |
C3H5Br | |
MFCD00000244 | |
allyl bromide, 3-bromopropene, 1-propene, 3-bromo, 3-bromopropylene, 3-bromo-1-propene, bromallylene, 2-propenyl bromide, 1-bromo-2-propene, propene, 3-bromo, allylbromide | |
3-bromoprop-1-ene |
Specifications
106-95-6, 75-56-9 | |
100.0 | |
Colorless | |
70.0°C to 71.0°C | |
Authentic | |
The liquid has a pungent odor | |
H2C=CHCH2Br | |
MFCD00000244 | |
13,219 | |
1.39 | |
Solubility in water: insoluble. Other solubilities: miscible with alcohol, chloroform, ether,, carbontetrachloride and carbon disulfide | |
C=CCBr | |
300ppm propylene oxide | |
7841 | |
99% | |
Allyl bromide, Stabilized |
98.5 | |
-119.0°C | |
1.3900g/mL | |
−1°C | |
98.5% min. (GC) | |
C3H5Br | |
1.4680 to 1.4700 | |
01,201 | |
15,279 | |
allyl bromide, 3-bromopropene, 1-propene, 3-bromo, 3-bromopropylene, 3-bromo-1-propene, bromallylene, 2-propenyl bromide, 1-bromo-2-propene, propene, 3-bromo, allylbromide | |
BHELZAPQIKSEDF-UHFFFAOYSA-N | |
3-bromoprop-1-ene | |
120.98 | |
120.98 | |
Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Toxic if inhaled.
Toxic if swallowed.
Very toxic to aquatic life.
Highly flammable liquid and vapor.
May cause genetic defects.
May cause cancer.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-446-6
- Allyl bromide, stabilized
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- ACUTE AQUATIC Acute 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H314-Causes severe skin burns and eye damage.
- H331-Toxic if inhaled.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Allyl bromide, Propylene oxide
RUO – Research Use Only