missing translation for 'onlineSavingsMsg'
Learn More
Allyl iodide, 97+%, stab. with copper, Thermo Scientific Chemicals Product Code.: 11448980

Allyl iodide, 97+%, stab. with copper, Thermo Scientific Chemicals

Product Code. 11448980
50 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11448980

Brand: Thermo Scientific Alfa Aesar A11822.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Allyl iodide is used as a reagent used with allylindium sesquiiodide in the cis-double allylation of cyclopropenes. It is used in synthesis of other organic compounds such as N-alkyl 2-pyrrolidone, Sorbic acid ester etc,.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyl iodide is used as a reagent used with allylindium sesquiiodide in the cis-double allylation of cyclopropenes. It is used in synthesis of other organic compounds such as N-alkyl 2-pyrrolidone, Sorbic acid ester etc,.

Solubility
Soluble in alcohol, chloroform, ether, and diethyl ether. Insoluble in water.

Notes
Store at 4°C. Protect from heat. Store away from oxidizing agents. Incompatible with heat, light, amines, nitrides, azo/diazo compounds, alkali metals, and epoxides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-56-9
Molecular Formula C3H5I
Molecular Weight (g/mol) 167.98
MDL Number MFCD00001094
InChI Key HFEHLDPGIKPNKL-UHFFFAOYSA-N
Synonym allyl iodide, 1-propene, 3-iodo, 3-iodopropene, 3-iodo-1-propene, 3-iodopropylene, propene, 3-iodo, allyliodide, unii-46830qoa4d, 3-iodo-propene, 3-iodanylprop-1-eneShow MoreShow Less
PubChem CID 11166
IUPAC Name 3-iodoprop-1-ene
SMILES ICC=C

Specifications

Melting Point -99°C
Density 1.832
Boiling Point 101°C to 103°C
Flash Point 18°C (64°F)
Linear Formula CH2=CHCH2I
Refractive Index 1.554
Quantity 50 g
UN Number UN1723
Beilstein 1697594
Sensitivity Light sensitive
Merck Index 14,294
Solubility Information Soluble in alcohol,chloroform,ether,and diethyl ether. Insoluble in water.
Formula Weight 167.98
Percent Purity ≥97%
Chemical Name or Material Allyl iodide, Stabilized with copper
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP26L009 Certificate TypeCertificate of Analysis Date28/03/2025 Catalog NumberA11822.30,A11822.18
Lot NumberM09L020 Certificate TypeCertificate of Analysis Date13/01/2025 Catalog NumberA11822.18
Lot NumberA0462073 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA11822.18
Lot NumberU08K034 Certificate TypeCertificate of Analysis Date14/08/2024 Catalog NumberA11822.18
Lot NumberU23J005 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog NumberA11822.30,A11822.18
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Allyl iodide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H300-Fatal if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Allyl iodide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Allyl iodide, 97+%, stab. with copper, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.