missing translation for 'onlineSavingsMsg'
Learn More
(+)-alpha-Pinene, 98%, Thermo Scientific Chemicals Product Code.: 11311777

(+)-alpha-Pinene, 98%, Thermo Scientific Chemicals

Product Code. 11311777
250 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11311777

Brand: Thermo Scientific Alfa Aesar L04941.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

(+)-alpha-Pinene is employed in the preparation of chiral hydroboration reagents. It is a bronchodilator in humans. α-Pinene is an anti-inflammatory and used as a broad-spectrum antibiotic.It exhibits activity as an acetylcholinesterase inhibitor, aiding memory. It is a biosynthetic base for CB2 ligands, such as HU-308.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+)-alpha-Pinene is employed in the preparation of chiral hydroboration reagents. It is a bronchodilator in humans. α-Pinene is an anti-inflammatory and used as a broad-spectrum antibiotic.It exhibits activity as an acetylcholinesterase inhibitor, aiding memory. It is a biosynthetic base for CB2 ligands, such as HU-308.

Solubility
Soluble in Ether, Alcohols, Chloroform. Not miscible in water.

Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7785-70-8
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00001346
InChI Key GRWFGVWFFZKLTI-RKDXNWHRSA-N
Synonym +-alpha-pinene, 1r,5r-2,6,6-trimethylbicyclo 3.1.1 hept-2-ene, 1r-+-alpha-pinene, 1r,5r-alpha-pinene, unii-h6cm4twh1w, +-pinene, h6cm4twh1w, 1r,5r-2-pinene, bicyclo 3.1.1 hept-2-ene, 2,6,6-trimethyl-, 1r,5r, +-pin-2 3-eneShow MoreShow Less
PubChem CID 82227
ChEBI CHEBI:28261
IUPAC Name (1R,5R)-4,6,6-trimethylbicyclo[3.1.1]hept-3-ene
SMILES CC1=CC[C@@H]2C[C@H]1C2(C)C

Specifications

Melting Point -62°C
Density 0.858
Boiling Point 154°C to 156°C
Flash Point 32°C (89°F)
Refractive Index 1.466
Quantity 250 g
UN Number UN2368
Beilstein 3648264
Merck Index 14,7445
Solubility Information Soluble in Ether,Alcohols,Chloroform. Not miscible in water.
Formula Weight 136.24
Percent Purity 98%
Chemical Name or Material (+)-alpha-Pinene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252386 Certificate TypeCertificate of Analysis Date18/09/2024 Catalog NumberL04941.30,L04941.18
Lot Number10249183 Certificate TypeCertificate of Analysis Date25/12/2023 Catalog NumberL04941.30,L04941.18
Lot Number10248701 Certificate TypeCertificate of Analysis Date22/11/2023 Catalog NumberL04941.30,L04941.18
Lot Number10220225 Certificate TypeCertificate of Analysis Date11/10/2023 Catalog NumberL04941.30,L04941.18
Lot Number10236369 Certificate TypeCertificate of Analysis Date11/10/2023 Catalog NumberL04941.30,L04941.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(+)-alpha-Pinene, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.