missing translation for 'onlineSavingsMsg'
Learn More
Aluminum Chloride Hydrate, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical Product Code.: 10447820

Aluminum Chloride Hydrate, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical

Product Code. 10447820
500g, Plastic powder jar
Click to view available options
500g, Plastic powder jar
This item is not returnable. View return policy

Product Code. 10447820

Brand: Fisher Chemical A/1960/53

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

500g, Plastic powder jar

TRUSTED_SUSTAINABILITY

Specifications

CAS 7784-13-6
Color White
pH 2.5 to 3.5
Physical Form Solid
Quantity 500 g
Molecular Formula AlCl3H12O6
MDL Number 149134
InChI Key JGDITNMASUZKPW-UHFFFAOYSA-K
SMILES O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]
IUPAC Name aluminium(3+) hexahydrate trichloride
Molecular Weight (g/mol) 241.42
Formula Weight 241.43g/mol
Packaging Plastic powder jar
Vapor Pressure 1mmHg 100
Show More Show Less
compliance-icons
Product Identifier
  • Aluminium chloride hexahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash face, hands and anyA§ thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum Chloride Hydrate, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.