missing translation for 'onlineSavingsMsg'
Learn More
Aluminum Sulfate Hexadecahydrate, Extra Pure, SLR, Fisher Chemical™ Product Code.: 10467820

Aluminum Sulfate Hexadecahydrate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10467820
5 kg, Plastic keg
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
This item is not returnable. View return policy

Product Code. 10467820

Brand: Fisher Chemical A/2560/65

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 16828-11-8
Molecular Formula Al2H22O20S3
Molecular Weight (g/mol) 492.30
MDL Number 149137
InChI Key JANHUGNIVASISO-UHFFFAOYSA-N
IUPAC Name tris(sulfuric acid) dialuminium octahydrate
SMILES O.O.O.O.O.O.O.O.[Al].[Al].OS(O)(=O)=O.OS(O)(=O)=O.OS(O)(=O)=O

Specifications

Melting Point 86°C
Color White
pH 2.5
Physical Form Solid
Quantity 5 kg
Formula Weight 342.1358g/mol
Packaging Plastic keg
compliance-icons
Product Identifier
  • Aluminium sulfate hexadecahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum Sulfate Hexadecahydrate, Extra Pure, SLR, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis